VersaTREK™ REDOX™ EZ Draw™ Media
Actual product may vary
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
VersaTREK™ REDOX™ EZ Draw™ Media
Thermo Scientific™
VersaTREK™ REDOX™ EZ Draw™ Media
Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, using the only FDA-cleared blood culture bottles for true direct draws.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeMedia TypeQuantity
7110-44MMicrobiology MediaAnaerobic manual media40 mL
7106-44Microbiology MediaAerobic media40 mL
7107-44Microbiology MediaAnaerobic media40 mL
Catalog number 7110-44M
Price (INR)
-
Product Type:
Microbiology Media
Media Type:
Anaerobic manual media
Quantity:
40 mL
Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, with Thermo Scientific™ VersaTREK™ REDOX™ EZ Draw™ Media, the only FDA-cleared true, direct measured draw in the industry. Accommodating up to 5mL of blood, EZ Draw media is available in a 40mL broth for both REDOX 1 aerobic and REDOX 2 anaerobic bottles and can be used for manual or automated monitoring of direct-draw blood samples. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the fully automated VersaTREK System's ability to better recover fastidious organisms.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Easy and convenient sampling
  • Reduced risk from sharps by eliminating the need for syringe, butterfly, angel wings and other blood transfer devices
  • Reduced costs by eliminating the need for separate collection devices
  • Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
  • Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
  • Available in 40 and 80mL sizes and suitable for all patient populations
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Manual/AutomatedManual
Media TypeAnaerobic manual media
No. per Pack50 Bottles/Box
Quantity40 mL
Specimen TypeBlood, Sterile Body Fluid
Volume (Metric) Specimen0.1 to 5 mL
Product TypeMicrobiology Media
Target Organism ClassAnaerobes
Test TypeMicrobial Detection
Unit SizeEach
Contents & Storage
Store away from direct sunlight