Product Image
Applied Biosystems™

Aquapore RP-18 7 μm, 0.3 x 1.0 cm (for the 140C instrument)

Have Questions?
Catalog NumberQuantity
7110221
also known as 711-0221
1 unit
Catalog number 7110221
also known as 711-0221
Price (INR)
37,427.00
Each
Quantity:
1 unit
Price (INR)
37,427.00
Each
This is a replacement part for the 140C Instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 unit
Shipping ConditionRoom Temperature
Product TypeAquapore RP-18
Unit SizeEach
Contents & Storage
Store at room temperature.