VersaTREK™ Myco Growth Supplement (Myco GS)
VersaTREK™ Myco Growth Supplement (Myco GS)
VersaTREK™ Myco Growth Supplement (Myco GS)
Thermo Scientific™
VersaTREK™ Myco Growth Supplement (Myco GS)
Promote growth of slow-growing mycobacteria within an inoculated bottle and facilitate detection with Thermo Scientific™ VersaTREK™ Myco Growth Supplement (Myco GS).
Have Questions?
Catalog NumberPackaging TypeMedia TypeQuantity
7112-4250 mL EachMyco growth supplement50 mL
Catalog number 7112-42
Price (INR)
-
Packaging Type:
50 mL Each
Media Type:
Myco growth supplement
Quantity:
50 mL
Promote growth of slow-growing mycobacteria within an inoculated bottle for enhanced detection with Thermo Scientific™ VersaTREK™ Myco Growth Supplement (Myco GS).
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionVersaTREK Myco Growth Supplement (Myco GS)
Media TypeMyco growth supplement
No. per Pack5
Packaging Type50 mL Each
Quantity50 mL
Specimen TypeAll Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Product TypeMicrobiology Media
SpeciesMycobacterium tuberculosis
Target Organism ClassMycobacteria
Test TypeMicrobial Detection, Antimicrobial Susceptibility
Unit SizeEach
Contents & Storage
Must be stored in the dark