Nalgene™ Lab Carrier
Thermo Scientific™
Nalgene™ Lab Carrier
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers.
Have Questions?
Catalog NumberHolds
7131-0001PK
also known as 7131-0001
Bottles
Catalog number 7131-0001PK
also known as 7131-0001
Price (INR)
14,199.00
Each
Holds:
Bottles
Price (INR)
14,199.00
Each
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers. The carrier used without inserts accommodates square and rectangular bottles or Nalgene™ Unwire™ test tube racks.
  • Remove sleeves to accommodate up to six bottles with 2.375 in. (6cm), 3.5 in. (9cm), or 4.25 in. (10.8cm) diameters
  • Sturdy, high density polyethylene construction holds six full 1000mL glass media bottles
  • Carriers stack when empty
Specifications
DescriptionLab Carrier
Diameter (Metric) Inner108 mm
No. per Case2
No. per Pack1
ColorWhite
HoldsBottles
MaterialPolyethylene
Positions6
Product TypeBottle Carrier
ShapeSquare
StackableYes
Unit SizeEach