para-JEM™ Reagent Kit w/ BLUE
para-JEM™ Reagent Kit w/ BLUE
para-JEM™ Reagent Kit w/ BLUE
Thermo Scientific™
para-JEM™ Reagent Kit w/ BLUE
Stock what's needed for faster detection of the causative agent of Johne's Disease on the Thermo Scientific™ VersaTREK™ System with a reagent kit including BLUE indicator dye.
Have Questions?
Catalog NumberQuantity
7137-42250 Tests/Kit
Catalog number 7137-42
Price (INR)
-
Quantity:
250 Tests/Kit
Achieve faster detection of M. avium ssp. paratuberculosis with the fully automated Thermo Scientific™ VersaTREK™ System and Thermo Scientific™ para-JEM™ Reagent Kit w/ BLUE. Kit includes para-JEM Broth, para-JEM Growth Supplement, para-JEM Antibiotic Supplement, para-JEM BLUE indicator dye and connectors for 250 tests on the VersaTREK System.
Solid medium culture used to be the standard procedure for detecting M. avium ssp. paratuberculosis (MAP), the causative agent of Johne's Disease, in ruminant fecal samples before para-JEM reagents. Due to the bacterium's long generation time, it can take up to 12 weeks of incubation on solid medium before the bacteria are detected. The para-JEM System is a unique, liquid culture method that combines a full line of reagents with the powerful Thermo Scientific™ VersaTREK™ Automated Microbial Detection System for faster, automated detection of MAP.
The para-JEM System includes a number of unique features:
  • Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
  • Provides increased sensitivity over conventional methods*
  • Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
  • Reduces labor with fewer false positives, false negatives and contaminants*
  • Improves workflow with most tests concluded at six weeks for high, medium and many low shedders*
Utilize para-JEM reagents with the fully automated VersaTREK Microbial Detection System, which automatically incubates and continuously monitors culture bottles for accurate, reliable organism detection as well as enhanced ergonomics and workflow versatility. Includes para-JEM Broth, para-JEM Growth Supplement, para-JEM Antibiotic Supplement and connectors.
Not all products are available for sale in all territories. Please inquire.
*Cited as a conclusion from an abstract presented at the 43rd Annual AAVLD Meeting - A New Liquid Culture Method, the TREK ESP Culture System II, for the Rapid Detection of Mycobacterium avium subsp. paratuberculosis in Bovine Fecal Samples, S.J.Shin, et. al, College of Veterinary Medicine Diagnostic Laboratory, Cornell University
Specifications
Descriptionpara-JEM Reagent Kit w/ BLUE
Packaging Type250 Tests/Kit
Quantity250 Tests/Kit
Specimen TypeVarious
Product Typepara-JAM Microbial Detection
SpeciesMycobacterium avium subsp. paratuberculosis
Test TypeMicrobial Detection
Unit SizeEach