Product Image
Thermo Scientific™

MPLC Seals (for 140C instrument)

Have Questions?
Catalog NumberQuantity
7150024
also known as 715-0024
1 Unit
Catalog number 7150024
also known as 715-0024
Price (INR)
-
Quantity:
1 Unit
This is a replacement part for the 140C Instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 Unit
Shipping ConditionRoom Temperature
Product TypeMPLC Seal
Unit Size1 unit
Contents & Storage
Store at room temperature.