Product Image
Invitrogen™

Anti-Mouse/Rat Foxp3 Staining Set PE, eBioscience™

The FJK-16s antibody reacts with mouse, rat, dog, porcine, and bovine Foxp3 also known as FORKHEAD BOX P3, SCURFIN, andRead more
Have Questions?
Catalog NumberQuantity
72-5775-401 kit
Catalog number 72-5775-40
Price (INR)
54,705.00
Each
Quantity:
1 kit
Price (INR)
54,705.00
Each
The FJK-16s antibody reacts with mouse, rat, dog, porcine, and bovine Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3 a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.

Intracellular staining of mouse splenocytes with FJK-16s using the PE anti-mouse/rat Foxp3 Staining Set and protocol reveals approximately 2% of total cells in the C57Bl/6 strain and approximately 3-5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+CD25+ cells and 4% of the CD4+CD25- cells staining with FJK-16s. B220+, CD11b+, CD11c+, and Ly6G/Gr-1+ cells do not show significant co-staining with FJK-16s.

Please see our FAQ regarding the usage of eBioscience Foxp3 reagents.

Not included:
Fc Block (cat. 14-0161)
Flow Cytometry Staining Buffer (cat. 00-4222)
Rat IgG2a isotype Control (cat. 12-4321, 11-4321, or 17-4321)

Isotype
IgG2a, kappa

Laser
Blue Laser

Emit
575 nm

Excite
488 nm

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenFoxp3
CloneFJK-16s
Concentration0.2mg/mL
Primary or SecondaryPrimary
Product LineeBioscience
ApplicationsFlow Cytometry
ClassificationMonoclonal
ConjugatePE
Host SpeciesRat
IsotypeIgG2a κ
Quantity1 kit
Regulatory StatusRUO
Target SpeciesMouse, Rat
Unit SizeEach
Contents & Storage
Store at 2-8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.