CO<sub>2</sub> and LN<sub>2</sub> Backup Systems
Actual product may vary
Thermo Scientific™
CO2 and LN2 Backup Systems
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems with ultra low-temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeStyle
7218TS
also known as 7218
Revco Value SI (11 cu. ft.)CO2 Backup SystemFreestanding
6214Thermo Scientific -80°C Chest FreezersLN2 Backup SystemFreestanding
6595Thermo Scientific Cryogenic FreezersLN2 Backup SystemBuilt-in
7216Revco PLUS HD (31.4 cu. ft.)CO2 Backup System, Factory-InstalledBuilt-in
6595-LN2PRevco PLUS, Revco PLUS HD (except 31.4, 11 cu. ft.)LN2 Backup SystemBuilt-in
6593DThermo Scientific -80°C Chest Freezers (208V)CO2 Backup SystemFreestanding
7219-LN2PRevco Value SI (11 cu. ft.)LN2 Backup SystemFreestanding
7215-LN2PRevco PLUS HD (31.4 cu. ft.)LN2 Backup SystemFreestanding
6594DRevco PLUS HD (except 31.4, 11 cu. ft.) (208V)CO2 Backup SystemBuilt-in
6214-LN2PRevco PLUS, Revco PLUS HD (except 31.4, 11 cu. ft.)LN2 Backup SystemFreestanding
6593-CO2PRevco PLUS, Revco PLUS HD (except 31.4, 11 cu. ft.)CO2 Backup SystemFreestanding
7214-CO2PRevco PLUS HD (31.4 cu. ft.)CO2 Backup System, Field-installed
6594-CO2PRevco PLUS, Revco PLUS HD (except 31.4, 11 cu. ft.), Revco™ PLUS HD (except 31.4, 11 cu. ft.)CO2 Backup System, Factory-InstalledFreestanding
6593AThermo Scientific -80°C Chest Freezers (115V)CO2 Backup SystemFreestanding
6594ARevco PLUS HD (except 31.4, 11 cu. ft.) (115V), Revco™ PLUS, Revco™ PLUSCO2 Backup SystemBuilt-in
Catalog number 7218TS
also known as 7218
Price (INR)
-
For Use With (Equipment):
Revco Value SI (11 cu. ft.)
Type:
CO2 Backup System
Style:
Freestanding
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems with ultra low-temperature freezers. Systems inject liquid CO2 or LN2 into cabinet when cabinet temperature warms to preset level.
Features
  • Temperature is adjustable in 1°C increments to -75°C (CO2), -150°C (LN2).
  • Digital display constantly indicates actual cabinet temperature.
  • Indicator lights come on when system is activated.
  • Additional indicator lights when cylinder is empty.
  • Liquid flow is shut off automatically when freezer door or lid is opened.
  • System is powered by rechargeable battery.
  • Low-battery light indicates when battery requires charging.
Ordering Information:
Thermo Scientific Revco™ Ultima™ II and Elite™ 3.1 cu. ft. (88L) models and all Value series models require freestanding system; only Ultima II and Elite series models larger than 3.1 cu. ft. (88L) accept built-in systems. Specify freezer model and voltage when ordering.
Note:
Customer-installed units; must be installed by qualified professional.
Includes:
All required hardware except CO2 or LN2 cylinder (must be ordered separately)
Specifications
DefrostAutomatic
For Use With (Equipment)Revco Value SI (11 cu. ft.)
TypeCO2 Backup System
Capacity (English)3.1 cu. ft.
Capacity (Metric)88 L
StyleFreestanding
Temperature Range-150°C to +1°C
Unit SizeEach