Manostat™ Silicone Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Silicone Pumping Links
Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeProduct Type
7230014514Pumping Link
7230025525Pumping Link
7230017517Pumping Link
7530013513Pumping Link
7230013513Pumping Link
7230016516Pumping Link
7530014514Pumping Link
7530016516Pumping Link
7530025525Pumping Link
7530017517Pumping Link
Catalog number 72300145
Price (INR)
-
Tubing Size:
14
Product Type:
Pumping Link
Thermo Scientific™ Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Specifications
For Use WithSimon/Vera, Kate and Compulab RD2
Product LineThermo Scientific
Tubing Size14
Diameter (English) Inner1/16 in.
Diameter (English) Outer0.55 in.
Diameter (Metric) Inner1.6 mm
Diameter (Metric) Outer14 mm
MaterialSilicone
Product TypePumping Link
Unit SizeEach