Manostat™ Norprene™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Norprene™ Pumping Links
Manostat norprene pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeDescription
7230525525Simon/Vera, Kate and Compulab RD2
7530525525Preston, Spencer and Compulab RD1
7530514514Preston, Spencer and Compulab RD1
7530516516Preston, Spencer and Compulab RD1
7530513513Preston, Spencer and Compulab RD1
7530517517Preston, Spencer and Compulab RD1
7230514514Simon/Vera, Kate and Compulab RD2
7230513513Simon/Vera, Kate and Compulab RD2
7230516516Simon/Vera, Kate and Compulab RD2
7230517517Simon/Vera, Kate and Compulab RD2
Catalog number 72305255
Price (INR)
-
Tubing Size:
25
Description:
Simon/Vera, Kate and Compulab RD2
Thermo Scientific™ Manostat norprene pumping links are for use with Manostat pumps and dispensers.
Specifications
DescriptionSimon/Vera, Kate and Compulab RD2
For Use WithSimon/Vera, Kate and Compulab RD2
Includes5 links/pack
Product LineThermo Scientific
Tubing Size25
Diameter (English) Inner3/16 in.
Diameter (English) Outer0.975 in.
Diameter (Metric) Inner4.8 mm
Diameter (Metric) Outer25 mm
MaterialNorprene
Product TypePumping Link
Unit SizeEach