Search
Search
| Catalog Number | Volume (Metric) | Sterility | Graduations | Packaging Type |
|---|---|---|---|---|
| 728NL | 13.7 mL | Non-sterile | 1, 2, 3, 4, 5 mL | Bulk 250/Pk 10Pk/Cs 2500/Cs |
| 274 | 4.6 mL | Non-sterile | 0.5, 1, 1.5, 2 mL | Bulk 500/Pk 10Pk/Cs 5000/Cs |
| 282TS also known as 282 | 1.5 mL | Non-sterile | 0.1 , 0.2, 0.3 mL | Bulk 500/Pk 8Pk/Cs 4000/Cs |
| 282-1S | 1.5 mL | Sterile | 0.1, 0.2, 0.3 mL | Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs |
| 718TS also known as 718 | 5.8 mL | Non-sterile | 0.25, 0.5, 0.75, 1 mL | Bulk 500/Pk 10Pk/Cs 5000/Cs |
| 222-1S | 5.8 mL | Sterile | 0.25, 0.5, 0.75, 1 mL |