Product Image
Invitrogen™

Anti-Human Foxp3 Staining Set Alexa Fluor™ 488 , eBioscience™

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. TheRead more
Have Questions?
Catalog NumberQuantity
73-5776-401 kit
Catalog number 73-5776-40
Price (INR)
41,532.00
Each
Quantity:
1 kit
Price (INR)
41,532.00
Each
This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The PCH101 monoclonal antibody reacts with the amino terminus of human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The PCH101 antibody crossreacts with rhesus, chimpanzee, and cynomolgus Foxp3 PCH101 recognizes a different epitope of Foxp3 than clones 236A/E7 and 150D/E4.

Isotype
IgG2a, kappa

Laser
Blue Laser

Emit
519 nm

Excite
488 nm

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenFoxp3
ClonePCH101
Gene AliasForkhead Box P3, Scurfin, JM2
Primary or SecondaryPrimary
Product LineeBioscience
ApplicationsFlow Cytometry
ClassificationMonoclonal
ConjugateAlexa Fluor 488
Host SpeciesRat
IsotypeIgG2a κ
Quantity1 kit
Regulatory StatusRUO
Target SpeciesChimpanzee, Cynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
Unit SizeEach
Contents & Storage
Store at 2-8°C. Do not freeze. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.