CaptureSelect™ HRP Anti-VSVG Conjugate
Thermo Scientific™

CaptureSelect™ HRP Anti-VSVG Conjugate

CaptureSelect HRP Anti-VSVG Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity and selectivity to the ectodomain of VSVG (the glycoprotein (G) of vesicular stomatitis virus).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7304072100100 μL
7304072500500 μL
Catalog number 7304072100
Price (INR)
64,819.00
100 μL
Quantity:
100 μL
Price (INR)
64,819.00
100 μL

CaptureSelect HRP Anti-VSVG Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity and selectivity to the ectodomain of VSVG (the glycoprotein (G) of vesicular stomatitis virus).

The affinity ligand is chemically conjugated to horseradish peroxidase (HRP) via an appropriate spacer that fully retains the binding reactivity of the ligand. This HRP conjugate enables the detection of free VSVG protein and membrane-bound VSVG, as displayed on vesicular stomatitis virus or VSVG pseudo-typed viruses and extracellular vesicles (EVs).

To set up a sensitive sandwich ELISA to quantitate free VSVG protein and VSVG-bearing particles, we recommend the use of CaptureSelect HRP Anti-VSVG Conjugate as detection reagent in combination with CaptureSelect Biotin Anti-VSVG Conjugate (Cat. No. 7104082100). This Biotin Anti-VSVG conjugate targets a different, non-competitive epitope on VSVG and can easily be immobilized onto streptavidin-coated surfaces to serve as a capture reagent in ELISA.

Product details

Format type: HRP, liquid

Buffer: PBS, 50% (v/v) glycerol

Storage: in freezer (-5°C to -30°C)

Concentration: 0.5 mg/mL

Volume: 0.1 mL

Recommended working dilution for sandwich ELISA: 1: 2,000 to 1: 10,000 (the optimal working dilution should be determined empirically for each application)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.5 mg/mL
FormulationPBS (pH 7.4), 50% glycerol
Quantity100 μL
Shipping ConditionDry ice
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit Size100 μL
Contents & Storage
Store in freezer (-30°C to -5°C).