Product Image
Applied Biosystems™
CT Supplement
CT Supplement
Have Questions?
Catalog NumberQuantity
740011000 tests
Catalog number 74001
Price (INR)
13,960.00
Each
Quantity:
1000 tests
Price (INR)
13,960.00
Each
CT Supplement is added to standard Sorbitol MacConkey media (SMAC) to form CT-SMAC. CT-SMAC selects for E.coli 0157 after IMS.
This Product Contains: 1mg Cefixime powder and 50mg Potassium Tellurite powder.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLiquid Supplement
Media TypePathogen Detection Media
Preparation MethodPrepare Stock Solution
Quantity1000 tests
Shipping ConditionRoom Temperature
Strain DesignationsE. coli O157
FormPowder
Target Organism ClassE. coli
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).