TN-1065 Neo Angle Rotor
Thermo Scientific™
TN-1065 Neo Angle Rotor
Optimized for CsCl Plasmid DNA separations, the Thermo Scientific™ TN-1065 Neo Angle Rotor provides a very steep angle, supporting up to ten 12mL ultracentrifuge tubes.
Have Questions?
Catalog NumberCapacity
7500303410 x 12 mL
Catalog number 75003034
Price (INR)
-
Capacity:
10 x 12 mL
Optimized for CsCl Plasmid DNA separations, the Thermo Scientific™ TN-1065 Neo Angle Rotor provides a very steep angle, supporting up to ten 12mL ultracentrifuge tubes.
  • Supports performance up to 65,000rpm for 12mL ultracentrifuge tubes
  • Compatible with Thermo Scientific™ Sorvall™ WX+ Ultracentrifuge Series
Specifications
Angle10°
DescriptionTN-1065 Neo Angle Rotor
For Use With (Equipment)Thermo Scientific Sorvall WX+ UltraCentrifuge Series
K Factor23
Max. Tube Diameter16 mm
Max. Tube Length78 mm
Capacity10 x 12 mL
MaterialTitanium
Max. RCF401,614 x g
Max. Speed65,000 rpm
TypeNeo Angle Rotor
Unit SizeEach