TN-1865 Neo Angle Rotor for WX+ Ultraspeed Centrifuges
Thermo Scientific™
TN-1865 Neo Angle Rotor for WX+ Ultraspeed Centrifuges
Optimized for CsCl Plasmid DNA separations, the Thermo Scientific TN-1865 Neo Angle Rotor provides a very steep angle, with an 18-place 5mL tube capacity.
Have Questions?
Catalog NumberCapacity
7500303518 x 5 mL
Catalog number 75003035
Price (INR)
-
Capacity:
18 x 5 mL
Optimized for CsCl Plasmid DNA separations, the Thermo Scientific™ TN-1865 Neo Angle Rotor provides a very steep angle, with an 18-place 5mL tube capacity.
  • Supports performance up to 65,000rpm for 5mL ultracentrifuge tubes
  • Compatible with Thermo Scientific™ Sorvall™ WX+ Ultracentrifuge Series
Specifications
Angle10°
DescriptionTN-1865 Neo Angle Rotor
For Use With (Equipment)Thermo Scientific Sorvall WX+ UltraCentrifuge Series
K Factor15
Max. Tube Diameter13 mm
Max. Tube Length53 mm
Capacity18 x 5 mL
MaterialTitanium
Max. RCF430,874 x g
Max. Speed65,000 rpm
TypeNeo Angle Rotor
Unit SizeEach