TX-200 Swinging Bucket Rotor
Thermo Scientific™
TX-200 Swinging Bucket Rotor
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75003659Bucket
75003658Swinging Bucket Rotor
75003660Bucket Lid
75003687Replacement O-ring
Catalog number 75003659
Price (INR)
-
Type:
Bucket
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor.
Ordering Information:
Round buckets (75-003-659), biocontainment lids (75-003-660) and replacement O-rings (75-003-687) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
DescriptionRound Buckets
For Use With (Equipment)TX-200 Rotor
QuantitySet of 4
BiocontainmentYes
TypeBucket
Unit SizeEach