Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Thermo Scientific™
Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Select adapters for 450-500mL blood bags in round single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
750038576/8 x 500 mL Triple or Quad Blood Bag
750038586/8 x 450 mL Single or Double Blood Bag
Catalog number 75003857
Price (INR)
-
Request A Quote
Capacity:
6/8 x 500 mL Triple or Quad Blood Bag
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor or, utilizing the same bucket system, upgrade to the HAEMAFlex 8 swinging bucket rotor.
Select adapters for 450–500mL Triple or Quad blood bags or 450mL Single or Double blood bags.
Specifications
Description450-500mL Triple or Quad Blood Bag Adapter (set of 2)
For Use With (Equipment)Single blood bag bucket with filter pack
Capacity6/8 x 500 mL Triple or Quad Blood Bag
TypeBlood Bag Adapter
Unit SizeEach