Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Thermo Scientific™
Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Select adapters for 450-500mL blood bags in round single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
750038586/8 x 450 mL Single or Double Blood Bag
750038576/8 x 500 mL Triple or Quad Blood Bag
Catalog number 75003858
Price (INR)
-
Capacity:
6/8 x 450 mL Single or Double Blood Bag
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor or, utilizing the same bucket system, upgrade to the HAEMAFlex 8 swinging bucket rotor.
Select adapters for 450–500mL Triple or Quad blood bags or 450mL Single or Double blood bags.
Specifications
Description450mL Single or Double Blood Bag Adapter (set of 2)
For Use With (Equipment)Single blood bag bucket with filter pack
Capacity6/8 x 450 mL Single or Double Blood Bag
TypeBlood Bag Adapter
Unit SizeEach