HAEMAFlex™ 6/8 Swinging Bucket Rotors
HAEMAFlex™ 6/8 Swinging Bucket Rotors
Thermo Scientific™
HAEMAFlex™ 6/8 Swinging Bucket Rotors
Expand individual bucket capacity up to 550mL blood bags with the Thermo Scientific™ HAEMAFlex™ 6/8 rotors for the new Thermo Scientific™ blood banking centrifuges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityMax. RCFMax. SpeedDescription
750038616 x 550 mL Blood Bags (6 x 1000 mL)7,295 x g5,000 rpmHAEMAFlex 6 Swinging Bucket Rotor
750038818 x 550 mL Blood Bags (8 x 1000 mL)7,144 x g4,600 rpmHAEMAFlex 8 Swinging Bucket Rotor
Catalog number 75003861
Price (INR)
-
Request A Quote
Capacity:
6 x 550 mL Blood Bags (6 x 1000 mL)
Max. RCF:
7,295 x g
Max. Speed:
5,000 rpm
Description:
HAEMAFlex 6 Swinging Bucket Rotor
Address your lab's regular- to medium-throughput blood processing needs with the Thermo Scientific™ HAEMAFlex™ 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the Thermo Scientific™ HAEMAFlex™ 8 swinging bucket rotor.
High-throughput sample processing with the flexibility to match your current and future needs with 6 or 8 x 550mL blood bag (6 or 8 x 1000mL) capacity, both available with the same bucket selections. Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
These windshielded rotors feature Thermo Scientific™ Eco-Spin™ technology, providing energy savings up to 64%, based on an engineering evaluation of windshielded and non-windshielded designs of the same rotor body.
Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ functions enabling instant rotor lid opening, storage and rotor identification to save time and protects the integrity of your samples.
Specifications
DescriptionHAEMAFlex 6 Swinging Bucket Rotor
For Use With (Equipment)Thermo Scientific Cryofuge 8 and 16 and Sorvall BP 8 and 16 Centrifuges
Capacity6 x 550 mL Blood Bags (6 x 1000 mL)
MaterialStainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
Max. RCF7,295 x g
Max. Speed5,000 rpm
TypeSwinging Bucket Rotor
Unit SizeEach