Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Thermo Scientific™
Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Round and conical buckets offered in sets of four
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeCapacity
75005707Bucket LidNot Applicable
75005703Bucket8 x 50 mL
Catalog number 75005707
Price (INR)
-
Product Type:
Bucket Lid
Capacity:
Not Applicable
Round and conical buckets or lids offered in sets of four
Specifications
CapacityNot Applicable
DescriptionClickSeal Biocontainment Lids for Round Buckets
For Use With (Equipment)Round Buckets
QuantitySet of 4
Product TypeBucket Lid
Unit SizeEach