LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Actual product may vary
Thermo Scientific™
LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Support a diverse array of sample processing applications without the need to modify existing protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75006479Bucket Lid
75006421Sealing Cap
75006436Double Blood Bag Bucket
75006476Swinging Bucket Rotor
75006477Bucket Rotor
75006478Double Spin Bucket
75006486Microplate Adapter
Catalog number 75006479
Price (INR)
-
Type:
Bucket Lid
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Cryofuge 5500i series centrifuges
Specifications
DescriptionBiocontainment Sealing Lid
For Use With (Equipment)Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
BiocontainmentYes
TypeBucket Lid
Unit SizeEach