Multifuge X1 R Pro, 208-240V TX-400 Cell Culture Package
Thermo Scientific™
Multifuge X1 R Pro, 208-240V TX-400 Cell Culture Package
Waste no time finding the right part numbers for your application! The application driven centrifuge package insures convinient, fast ordering of all components necessary to run a specific protocol.
Have Questions?
Catalog number 75016023
Price (INR)
-
Waste no time finding the right part numbers for your application! The application driven centrifuge package insures convinient, fast ordering of all components necessary to run a specific protocol. Run 50 mL, 15 mL conical tubes without changing rotors or buckets. Contains centrifuge, TX-400 rotor, buckets, biocontainment lids, 50 mL conical adapters (maximum capacity 16x50 mL), 15 mL conical adapter, (maximum capacity 36 x 15 mL).
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Drive SystemDirect, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
IncludesPart numbers in Package: 75009750 (centrifuge),75003181(rotor),75003655(set of buckets),75003656(lids),75003682(set of 15 mL adapters),75003683(set of 50 mL adapters)
Product TypeGeneral Purpose Centrifuge Series
Unit SizeEach