General Purpose Centrifuge Rotor Packages
Actual product may vary
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
Thermo Scientific™
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
75016109TX-750 Blood Tube Package
75016103TX-1000 Blood Tube Package
75016104TX-1000 Microplate Package
75016105TX-1000 Cell Culture Package
75016106TX-200 Cell Culture Package
75016107TX-400 Blood Tube Package
75016108TX-400 Cell Culture Package
75016110TX-750 Cell Culture Package
75016111TX-200 Blood Tube Package
Catalog number 75016109
Price (INR)
-
Model:
TX-750 Blood Tube Package
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
IncludesTX-750 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
ModelTX-750 Blood Tube Package
Net WeightSee individual components of package
WarrantySee Manual
BiocontainmentYes
CapacityMax in TX-750: 148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
MaterialSee individual centrifuge, rotor and accessory data
TypeRotor Equipment Package
Unit SizeEach