Manostat™ Silicone Pumping Links
Manostat™ Silicone Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Silicone Pumping Links
Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeProduct Type
7530025525Pumping Link
7230025525Pumping Link
7230017517Pumping Link
7530013513Pumping Link
7230013513Pumping Link
7230016516Pumping Link
7530014514Pumping Link
7530016516Pumping Link
7230014514Pumping Link
7530017517Pumping Link
Catalog number 75300255
Price (INR)
-
Request A Quote
Tubing Size:
25
Product Type:
Pumping Link
Thermo Scientific™ Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Specifications
For Use WithPreston, Spencer and Compulab RD1
Product LineThermo Scientific
Tubing Size25
Diameter (English) Inner3/16 in.
Diameter (English) Outer0.975 in.
Diameter (Metric) Inner4.8 mm
Diameter (Metric) Outer25 mm
MaterialSilicone
Product TypePumping Link
Unit SizeEach