Manostat™ Tygon™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Tygon™ Pumping Links
Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeDescription
7531014514Preston, Spencer and Compulab RD1
7231017517Simon/Vera, Kate, Compulab RD2
7531025525Preston, Spencer and Compulab RD1
7531016516Preston, Spencer and Compulab RD1
7531017517Preston, Spencer and Compulab RD1
7231013513Simon/Vera, Kate, Compulab RD2
7231025525Simon/Vera, Kate, Compulab RD2
7231016516Simon/Vera, Kate, Compulab RD2
7231014514Simon/Vera, Kate, Compulab RD2
7531013513Preston, Spencer and Compulab RD1
Catalog number 75310145
Price (INR)
-
Tubing Size:
14
Description:
Preston, Spencer and Compulab RD1
Thermo Scientific™ Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Specifications
DescriptionPreston, Spencer and Compulab RD1
For Use WithPreston, Spencer and Compulab RD1
Includes5 links/pack
Product LineThermo Scientific
Tubing Size14
Diameter (English) Inner1/16 in.
Diameter (English) Outer0.546 in.
Diameter (Metric) Inner1.6 mm
Diameter (Metric) Outer14 mm
MaterialTygon
Product TypePump Tubing Links
Unit SizeEach