Rheodyne™ 7725 and 7725i Sample Injectors
Actual product may vary
Rheodyne™ 7725 and 7725i Sample Injectors
Rheodyne™ 7725 and 7725i Sample Injectors
Thermo Scientific™
Rheodyne™ 7725 and 7725i Sample Injectors
Ensure accurate, precise HPLC injections and protect against pressure shock with the dual-mode Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
7725IContinuous Flow, Position Sensing Switch
7725Continuous Flow
Catalog number 7725I
Price (INR)
-
Description:
Continuous Flow, Position Sensing Switch
Maintain continuous HPCL flow between the load and inject positions and protect against pressure shock by using Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors. The Make-Before-Break (MBB) design ensures uninterrupted flow during sample injection. These stainless-steel, dual-mode injectors can use partial filling for zero sample waste or complete filling for better reproducibility. Samples from 1μL to 5mL can be injected with high accuracy and precision. The 7725i model features a position-sensing switch for a reproducible start signal.
  • Stainless steel construction
  • Make-Before-Break (MBB) design
  • Can use partial filling for zero sample waste or complete filling for better reproducibility
  • Inject 1μL to 5mL with high accuracy and precision
  • 7725i features a position sensing switch for a reproducible start signal
(Includes: 20μL sample loop)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionContinuous Flow, Position Sensing Switch
For Use With (Equipment)HPLC
ModeDual Mode
Stator MaterialStainless Steel
TypeContinuous Flow HPLC Sample Injector
Volume (Metric) Sample20 μL
Unit SizeEach