Product Image
Invitrogen™

MYR PKC Zeta Pseudosubstrate

MYR PKC Zeta Pseudosubstrate
Have Questions?
Catalog NumberQuantity
77749
also known as 77-749
1 mg
Catalog number 77749
also known as 77-749
Price (INR)
-
Quantity:
1 mg
Selective protein kinase C (PKC) zeta inhibitor.

Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.

Molecular Weight: 1928
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gene ID (Entrez)5590
Inhibitor PropertiesPeptide
InhibitorsKinase Inhibitor (PKC Inhibitor), Kinase Inhibitor
Molecular Weight (g/mol)1928 Da
Purification MethodHPLC
Quantity1 mg
Shipping ConditionDry Ice
Specific Inhibit or TargetPKCζ
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).