Search
| Catalog Number | Diameter (English) Inner | Diameter (English) Outer |
|---|---|---|
| 8051-0187PK also known as 8051-0187 | 1/8 in. | 3/16 in. |
| 8051-0125PK also known as 8051-0125 | 1/16 in. | 1/8 in. |
Utilize translucent Thermo Scientific™ Nalgene™ 870 Tubing for transferring aggressive and corrosive chemical fluids. Useful over a very wide temperature range (-270°C to +260°C) with low water absorption. Excellent for low-pressure use with mechanical, compression fittings at room tempearature. Food-grade materials comply with 21 CFR 177.1520 for food packaging.
Note:
Do not sterilize by radiation. Should not be used with molten alkali metals, elemental fluorine and fluorine precursors at elevated temperatures or concentrated perchloric acid.
Please recycle the corrugate cartons in which your Nalgene products are packaged.