CaptureSelect™ C1-InhXL Ligand Leakage ELISA
CaptureSelect™ C1-InhXL Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ C1-InhXL Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ C1-InhXL Ligand Leakage ELISA is designed to detect possible leached ligand of C1-esterase inhibitor (C1Inh) purified using POROS CaptureSelect C1-InhXL Affinity Resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81034021010 Assays
8103402011 Assay
Catalog number 810340210
Price (INR)
-
Quantity:
10 Assays
CaptureSelect C1-InhXL Ligand Leakage ELISA is designed to detect possible leached ligand of C1-esterase inhibitor (C1Inh) purified using POROS CaptureSelect C1-InhXL Affinity Resin. The C1-InhXL affinity ligand is a 12–15 kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the ligand can leach from the chromatography support and co-elute with the target protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect C1-InhXL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodELISA
FormatIndividual components
Quantity10 Assays
TargetCaptureSelect™ C1-InhXL Affinity Ligand
FormFrozen Liquid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).