CaptureSelect™ hCG Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ hCG Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ hCG Ligand Leakage ELISA is designed to detect possible leached ligand of human chorionic gonadotropin (hCG) purified using CaptureSelect hCG Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81034101010 Assay
8103410011 Assay
Catalog number 810341010
Price (INR)
1,194,050.00
Each
Quantity:
10 Assay
Price (INR)
1,194,050.00
Each
CaptureSelect hCG Ligand Leakage ELISA is designed to detect possible leached ligand of human chorionic gonadotropin (hCG) purified using CaptureSelect hCG Affinity Matrix. In this affinity matrix the hCG affinity ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The hCG Ligand Leakage ELISA kit can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microplate Reader
Product LineCaptureSelect
Quantity10 Assay
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
3 x 1 mL vials, store at -5°C to -30°C