CaptureSelect™ tPA Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ tPA Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ tPA Ligand Leakage ELISA is designed to detect possible leached ligand of tissue plasminogen activator (tPA) purified using CaptureSelect tPA Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81034301010 Assays
8103430011 Assay
Catalog number 810343010
Price (INR)
1,007,090.00
Each
Quantity:
10 Assays
Price (INR)
1,007,090.00
Each
CaptureSelect tPA Ligand Leakage ELISA is designed to detect possible leached ligand of tissue plasminogen activator (tPA) purified using CaptureSelect tPA Affinity Matrix. The ligand, a 13 kD recombinant protein, is covalently coupled to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect tPA Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 Assays
Shipping ConditionDry Ice
Product LineCaptureSelect
TypeELISA Kit
Unit SizeEach
Contents & Storage
3 x 1 mL, store at -5°C to -30°C.