CaptureSelect™ AAT-XL Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ AAT-XL Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81038321010 assay x 96-well plate
8103832011 assay x 96-well plate
Catalog number 810383210
Price (INR)
1,008,860.00
Each
Quantity:
10 assay x 96-well plate
Price (INR)
1,008,860.00
Each
CaptureSelect AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix. The AAT-XL affinity ligand is a 13-kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of ligand can leach from the chromatography support and co-elute with the target protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand concentrations greater than or equal to 1 ng/mL. The CaptureSelect AAT-XL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
For Use With (Application)Ligand Leakage ELISA assay
Quantity10 assay x 96-well plate
Shipping ConditionDry Ice
TargetTarget-specific ELISA reagents
FormFrozen Liquid
Product LineCaptureSelect
TypeELISA Kit
Unit SizeEach