Nalgene™ LDPE, Round Carboy with Spigot
Thermo Scientific™

Nalgene™ LDPE, Round Carboy with Spigot

For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Have Questions?
Catalog number 8318-0050PK
also known as 8318-0050
Price (INR)
-
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Features:
  • Molded in graduations of 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles that make lifting comfortable for four largest sizes
  • Molded-in spigot boss for strength and leak resistance
  • Leakproof†

Ordering Information:

Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.

Includes:

Polypropylene spigot and screw closure

Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.

Specifications
Capacity (English)5.3 gal.
Closure MaterialPolypropylene
Closure Size83B
DescriptionRound Carboys w/Spigot
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer286 mm
For Use With (Application)Collect and dispense distilled water, acids and reagents
GraduationsMolded-in graduations in 1 gal. and 5L increments
Height (Metric) With Closure528 mm
IncludesIntegral shoulder handles, molded-in spigot boss
No. per Case4/Cs.
No. per Pack1 Ea.
AutoclavableNot Autoclavable
Capacity (Metric)20 L
ColorWhite
MaterialLDPE
Product TypeCarboy with Spigot
Unit SizePack of 1