Nicolet™ Collection of FT-IR and Raman Matched Spectra
Thermo Scientific™

Nicolet™ Collection of FT-IR and Raman Matched Spectra

Designed for laboratories with both FT-IR and Raman spectra available, this collection of FT-IR and Raman Matched Spectra includes compounds of general interest.
Have Questions?
Catalog NumberType
833-009601FT-IR and Raman Matched Spectra
Catalog number 833-009601
Price (INR)
-
Type:
FT-IR and Raman Matched Spectra
Designed for laboratories with both spectra available, the Thermo Scientific™ Nicolet™ Collection of FT-IR and Raman Matched Spectra includes compounds of general interest.
This Nicolet Collection of FT-IR and Raman Matched Spectra contains a wide array of common chemicals found in The Aldrich Handbook of Fine Chemicals.
  • Contains 3,119 FT-IR spectra and 3,119 Raman spectra of general interest to the analytical laboratory customer
  • Contains a subset of the Aldrich FT-IR Condensed Phase Library along with matching Raman spectra of the same compounds
  • Combined searching requires OMNIC Linked Search software that is included with the package
  • Data format: Deresolved
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification — or at least classification — of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeFT-IR and Raman Matched Spectra
Unit SizeEach