Pharmaceutical Excipients Matched IR/Raman Library
Thermo Scientific™

Pharmaceutical Excipients Matched IR/Raman Library

Obtain high quality FT-IR and Raman spectra of common pharmaceutical excipients using the Thermo Scientific™ Pharmaceutical Excipients Matched IR/Raman Library.
Have Questions?
Catalog NumberType
833-023400Pharmaceutical Excipients Matched IR/Raman Library
Catalog number 833-023400
Price (INR)
-
Type:
Pharmaceutical Excipients Matched IR/Raman Library

Obtain high quality spectra of common pharmaceutical excipients using the Thermo Scientific™ Pharmaceutical Excipients Matched IR/Raman Library, designed for laboratories studying pharmaceutical formulations using both FT-IR and Raman.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

This library provides high quality FT-IR and Raman spectra of common pharmaceutical excipients.
  • Contains 300 matched FT-IR and Raman spectra
  • All materials were tested via current United States Pharmacopoeia compendium methodology for identity and purity—only materials that passed these methods were used to create this library
  • Created in collaboration with David E. Bugay and W. Paul Findlay of Bristol-Myers Squibb Pharmaceutical Research Institute
  • The spectra were collected using Thermo Scientific FT-IR and FT-Raman spectrometers
  • Also available individually: FT-IR library (Part No. 834-019201) and Raman library (Part No. 834-019301)
Includes:
Textbook with explanation of spectroscopy techniques and the spectra of pharmaceutical excipients for reference use.

About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypePharmaceutical Excipients Matched IR/Raman Library
Unit SizeEach