OMNIC™ Specta Software
OMNIC™ Specta Software
OMNIC™ Specta Software
Thermo Scientific™
OMNIC™ Specta Software
Quickly gather clear visual data as you work using the innovative identification and interpretation tools of the Thermo Scientific™ OMNIC™ Specta Software.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
833-036700Vapor Phase
833-036200FT-IR Standard
833-036300For Polymer Labs
833-036400For Forensic Labs
833-036500FT-IR Complete
833-036600For TGA
833-036800Standard Raman
833-036900Analytical Raman
833-043700Nicolet iS50 Edition
Catalog number 833-036700
Price (INR)
-
Type:
Vapor Phase
Quickly gather clear visual data as you work using the innovative identification and interpretation tools of Thermo Scientific™ OMNIC™ Specta Software.
OMNIC Specta Software creates a database of spectral data through is a unique combination of spectral identification tools, interpretation algorithms, and scientific documentation that indexes information on your hard disk. The software guides you in identifying materials and verifying assumptions—eliminating typical analytical tasks—so you get answers quickly and confidently.
The OMNIC Specta Software
  • Identify constituents in mixture samples with multi-component and contaminant searches — easier than tedious manipulations required by traditional search and subtract processing
  • Maintain historical data access to save time and prevent information loss 
  • Minimize time and effort with simple one click processing 
  • Receive immediate visual feedback for confidence in your results 
  • Ensure quality results with live displays for active data viewing 
  • Keep working during data collection for maximum productivity
  • Identify mixtures in one step for ultimate speed and confidence 
  • Communicate efficiently with modern PDF reports 
  • Copy directly into word processing programs
Specifications
For Use With (Application)Material Characterization
For Use With (Equipment)Nicolet iS10, iZ10 and 6700 FT-IR Spectrometers
TypeVapor Phase
Unit SizeEach