Aldrich™ Raman Condensed Phase Spectral Library
Aldrich™ Raman Condensed Phase Spectral Library
Thermo Scientific™

Aldrich™ Raman Condensed Phase Spectral Library

Increase efficiency in your lab with one of the most comprehensive collections of FT-Raman spectral references available.
Have Questions?
Catalog NumberType
834-004001Raman Condensed Phase Spectral Library
Catalog number 834-004001
Price (INR)
-
Request A Quote
Type:
Raman Condensed Phase Spectral Library

Increase efficiency in your lab with one of the most comprehensive collection of FT-Raman spectral references available. The Thermo Scientific™ Aldrich™ Raman Condensed Phase Spectral Library contains many common chemicals found in The Aldrich Handbook of Fine Chemicals.

Our comprehensive libraries take the guesswork out of your spectral analysis and feature fully-reviewed, high-quality spectroscopy databases—enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

To create the Aldrich Raman Condensed Phase Library, 14,033 compounds found in the Aldrich Collection of FT-IR Spectra Edition II Library were excited with an Nd:YVO4 laser (1064 nm) using laser powers between 400-600 mW, measured at the sample. A Thermo FT-Raman spectrometer (with a Ge detector) was used to collect the Raman spectra. The spectra were saved in Raman Shift format.
  • Contains 14,033 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeRaman Condensed Phase Spectral Library
Unit SizeEach