Sigma-Aldrich™ Sugars and Carbohydrates FT-IR Spectral Library
Thermo Scientific™

Sigma-Aldrich™ Sugars and Carbohydrates FT-IR Spectral Library

Confidently identify sugars and carbohydrates with this library, a material-specific subset of the larger Thermo Scientific™ Sigma Biochemical Condensed Phase Library.
Have Questions?
Catalog NumberType
834-005901Sugars and Carbohydrates FT-IR Spectral Library
Catalog number 834-005901
Price (INR)
-
Type:
Sugars and Carbohydrates FT-IR Spectral Library
Confidently identify sugars and carbohydrates with this library, a material-specific subset of the larger Thermo Scientific™ Sigma Biochemical Condensed Phase Library. The Thermo Scientific Sigma Sugars and Carbohydrates FT-IR Spectral Library features fully-reviewed, high-quality spectroscopy databases to confidently identify compounds, take the guesswork out of your spectral analysis and create greater efficiencies in your lab.
  • Spectra acquired by Sigma-Aldrich Co.™ and examined and processed at Thermo Fisher Scientific. The spectra include compound name, molecular formula, CAS (Chemical Abstract Service) registry number and Sigma catalog number.
  • Contains 614 spectra
  • Data format: Deresolved
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeSugars and Carbohydrates FT-IR Spectral Library
Unit SizeEach