Nicolet™ FT-IR Vapor Phase Spectral Library
Thermo Scientific™

Nicolet™ FT-IR Vapor Phase Spectral Library

Investigating gas phase materials? This spectral library offers one of the most comprehensive collections of vapor phase FT-IR spectra available.
Have Questions?
Catalog NumberType
834-006600Nicolet FT-IR Vapor Phase Spectral Library
Catalog number 834-006600
Price (INR)
-
Type:
Nicolet FT-IR Vapor Phase Spectral Library

Investigating gas phase materials? The Thermo Scientific™ Nicolet FT-IR Vapor Phase Spectral Library offers one of the most comprehensive collections of vapor phase FT-IR spectra available.

Our comprehensive libraries take the guesswork out of your spectral analysis and increase lab efficiency. They feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

This library is an indispensable tool for scientists investigating gas phase materials.
  • Contains FT-IR spectra of compounds measured in gas phase
  • Most spectra were acquired by the Sigma-Aldrich Company using product samples
  • Additional spectra were collected by Hannover University, University of Wurzburg and Thermo Fisher Scientific applications scientists
  • Contains 8,654 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeNicolet FT-IR Vapor Phase Spectral Library
Unit SizeEach