TGA Vapor Phase FT-IR Spectral Library
Thermo Scientific™

TGA Vapor Phase FT-IR Spectral Library

Study composition and decomposition properties with the aid of this FT-IR spectral library; includes spectra of compounds likely to evolve during TGA/FT-IR experiments.
Have Questions?
Catalog NumberType
834-007200TGA Vapor Phase FT-IR Spectral Library
Catalog number 834-007200
Price (INR)
-
Type:
TGA Vapor Phase FT-IR Spectral Library
Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. The Thermo Scientific™ TGA Vapor Phase FT-IR Spectral Library is a valuable tool for chemists studying composition and decomposition material properties, and includes spectra of compounds likely to evolve during TGA/FT-IR experiments.Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
This library contains FT-IR spectra of compounds commonly found when running TGA/FT-IR studies and will help identify most low molecular weight gases. However, investigators searching for spectra of more exotic species or for more combinations of functional groups should consider the Thermo Scientific™ Nicolet™ FT-IR Vapor Phase Library.
  • Contains 460 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeTGA Vapor Phase FT-IR Spectral Library
Unit SizeEach