Georgia State Crime Lab Drug FT-IR Spectral Library
Thermo Scientific™

Georgia State Crime Lab Drug FT-IR Spectral Library

Thermo Scientific™ Georgia State Crime Lab Drug FT-IR Spectral Library is one of the most extensively used collections for the identification of drug substances.
Have Questions?
Catalog NumberType
834-007501Georgia State Crime Lab Drug FT-IR Spectral Library
Catalog number 834-007501
Price (INR)
-
Type:
Georgia State Crime Lab Drug FT-IR Spectral Library

Thermo Scientific™ Georgia State Crime Lab Drug FT-IR Spectral Library is one of the most extensively used collections for the identification of drug substances.

This definitive forensic library contains FT-IR spectra of illegal and legal drugs as well as diluents, precursors and other drug-related compounds.
  • Consists of spectra that were acquired by the Georgia Bureau of Investigation
  • Samples were secured as pure drug standards from the U.S. Drug Enforcement Administration (DEA), Applied Science Laboratories, U.S. Pharmacopoeial Conventions, Inc., and various pharmaceutical companies
  • For the majority of samples the material purity is 95% or better and in many cases is greater than 99%
  • Unless otherwise stated, the samples were prepared for spectral analysis using KBr pellets
  • Contains 1940 spectra
  • Data format: High Resolution (HR)
Specifications
For Use With (Application)FT-IR
TypeGeorgia State Crime Lab Drug FT-IR Spectral Library
Unit SizeEach