Hummel Polymer and Additives FT-IR Spectral Library
Thermo Scientific™

Hummel Polymer and Additives FT-IR Spectral Library

Thermo Scientific™ Hummel Polymer and Additives FT-IR Spectral Library is one of the most comprehensive and commonly used libraries for analysis of polymer materials.
Have Questions?
Catalog NumberType
834-008601Hummel Polymer and Additives FT-IR Spectral Library
Catalog number 834-008601
Price (INR)
-
Type:
Hummel Polymer and Additives FT-IR Spectral Library

Thermo Scientific™ Hummel Polymer and Additives FT-IR Spectral Library is one of the most comprehensive and commonly used libraries for analysis of polymer materials.

This library includes spectra of polymers, solvents, monomers and industrial organic compounds and is a valuable resource for anyone involved in the identification of polymers, particularly those working in labs involved in polymer production or analysis. The Hummel Polymer and Additives Library contains spectra collected by Professor Dieter Hummel of the Institute of Physical Chemistry at the University of Cologne (Koln), a recognized specialist in polymer chemistry.
  • Designed to aid analysts in the identification of typical polymer impurities, including excess residual monomers and additives, and indicates the composition of completed products
  • Contains additional information such as manufacturer, trade name, literature reference and measurement conditions for each spectrum in the collection
  • Contains 2011 spectra
  • Data format: High Resolution (HR)
834-008601
Specifications
For Use With (Application)FT-IR
TypeHummel Polymer and Additives FT-IR Spectral Library
Unit SizeEach