Surfactants FT-IR Spectral Library
Thermo Scientific™

Surfactants FT-IR Spectral Library

Interested in surfactants and related materials? The Thermo Scientific™ Surfactants FT-IR Spectral Library offers a comprehensive collection.
Have Questions?
Catalog NumberType
834-009100Surfactants FT-IR Spectral Library
Catalog number 834-009100
Price (INR)
-
Type:
Surfactants FT-IR Spectral Library
Interested in surfactants and related materials? The Thermo Scientific™ Surfactants FT-IR Spectral Library offers a comprehensive collection to help take the guesswork out of spectral analysis and create greater efficiencies in your lab.Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
  • Designed for use by scientists with an interest in surfactants and related materials, this library's spectra were carefully characterized and compiled by Clara Craver of Chemir Analytical Services and are organized by trade name and formula.
  • Contains 637 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeSurfactants FT-IR Spectral Library
Unit SizeEach