Food Additives FT-IR Spectral Library
Thermo Scientific™
Food Additives FT-IR Spectral Library
Features condensed phase FT-IR spectra of compounds of interest to the food industry.
Have Questions?
Catalog NumberType
834-010300Food Additives FT-IR Spectral Library
Catalog number 834-010300
Price (INR)
-
Type:
Food Additives FT-IR Spectral Library
Thermo Scientific™ Food Additives FT-IR Spectral Library features condensed phase FT-IR spectra of compounds of interest to the food industry.
The included compounds are on the GRAS (Generally Recognized As Safe) list of US government-approved food additives.
  • Consists of 519 condensed phase FT-IR spectra
  • These spectra are complementary to the vapor phase spectra included in the Flavors and Fragrances Library (Product No. 834-009400)
  • Data format: High Resolution
Specifications
For Use With (Application)FT-IR
TypeFood Additives FT-IR Spectral Library
Unit SizeEach