Synthetic Fibers by Microscope FT-IR Spectral Library
Synthetic Fibers by Microscope FT-IR Spectral Library
Thermo Scientific™

Synthetic Fibers by Microscope FT-IR Spectral Library

Identify fibers of varying diameters from a wide variety of products and manufacturers with the Thermo Scientific™ Synthetic Fibers by Microscope FT-IR Spectral Library.
Have Questions?
Catalog NumberType
834-011100Synthetic Fibers - FT-IR Spectral Library
Catalog number 834-011100
Price (INR)
-
Request A Quote
Type:
Synthetic Fibers - FT-IR Spectral Library
Identify fibers of varying diameters from a wide variety of products and manufacturers with the Thermo Scientific™ Synthetic Fibers by Microscope FT-IR Spectral Library.Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
  • This library contains FT-IR microscope transmission spectra of synthetic fibers from the Collaborative Testing Services' Reference Collection of Synthetic Fibers. It consists of spectra obtained from a collaborative project between Thermo Fisher Scientific and Dr. Jay A. Siegel and his co-workers in the School of Criminal Justice at Michigan State University.
  • All spectra were measured from pressed samples of the fibers using a Thermo Scientific FT-IR spectrometer and microscope system.
  • Contains 76 spectra
  • Data format: Deresolved
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeSynthetic Fibers - FT-IR Spectral Library
Unit SizeEach