Hazardous Chemicals Condensed Phase Spectral Library
Hazardous Chemicals Condensed Phase Spectral Library
Thermo Scientific™

Hazardous Chemicals Condensed Phase Spectral Library

A collection of condensed phase spectra of toxic chemicals, pollutants and other contaminants that are of interest to scientists involved in hazardous waste site remediation.
Have Questions?
Catalog NumberType
834-011401Haz Chemicals Spectral Library
Catalog number 834-011401
Price (INR)
-
Request A Quote
Type:
Haz Chemicals Spectral Library

Thermo Scientific™ Hazardous Chemicals Condensed Phase Spectral Library contains a collection of condensed phase spectra of toxic chemicals, pollutants and other contaminants that are of interest to scientists involved in hazardous waste site remediation.

The Hazardous Chemicals Condensed Phase Library contains spectra of compounds found on the US Environmental Protection Agency's Office of Toxic Substances consolidated list of chemicals subject to reporting under Title III of the Superfund Amendments and Reauthorization Act of 1986 (SARA), also known as the Emergency Planning and Community Right-to-Know Act (EPCRA).
  • Contains 411 spectra
  • Data format: High Resolution (HR)
Specifications
For Use With (Application)FT-IR
TypeHaz Chemicals Spectral Library
Unit SizeEach