Paper Materials FT-IR Spectral Library
Thermo Scientific™

Paper Materials FT-IR Spectral Library

Determine the composition of completed paper products as well as identify impurities with the useful Thermo Scientific™ Paper Materials FT-IR Spectral Library.
Have Questions?
Catalog NumberType
834-013900Paper Materials FT-IR Spectral Library
Catalog number 834-013900
Price (INR)
-
Type:
Paper Materials FT-IR Spectral Library

Determine the composition of completed paper products as well as identify impurities with the useful Thermo Scientific™ Paper Materials FT-IR Spectral Library.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab with our comprehensive libraries, featuring fully-reviewed, high-quality spectroscopy databases for confident compound identification in a variety of environmental, forensic, pharmaceutical and industrial applications.

The spectra were chosen to aid in the identification of impurities as well as determining the composition of completed paper products.
  • Contains 302 spectra of compounds and materials used in the production of paper products
  • Spectra were collected using a variety of techniques, including transmission and ATR
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypePaper Materials FT-IR Spectral Library
Unit SizeEach