Raman Polymer Spectral Library
Thermo Scientific™
Raman Polymer Spectral Library
Need quick determination and identification of polymer packaging or contaminants? This library offers a selection of common industrial polymers, copolymers and additives.
Have Questions?
Catalog NumberType
834-014101Raman Polymer Spectral Library
Catalog number 834-014101
Price (INR)
-
Type:
Raman Polymer Spectral Library
Need quick determination and identification of polymer packaging or contaminants? The Thermo Scientific™ Raman Polymer Spectral Library conventiently offers a selection of common industrial polymers, copolymers and additives.Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
  • This library provides a collection of common industrial polymer compounds run using the Raman analytical technique, and includes spectra of polymers, copolymers and additives.
  • Includes 99 spectra of industrially important polymers
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeRaman Polymer Spectral Library
Unit SizeEach