Lubricants and Oils FT-IR Spectral Library
Lubricants and Oils FT-IR Spectral Library
Thermo Scientific™

Lubricants and Oils FT-IR Spectral Library

Consists of spectra representing an international collection of lubricants and oils.
Have Questions?
Catalog NumberType
834-016000Lubricants and Oils FT-IR Spectral Library
Catalog number 834-016000
Price (INR)
-
Request A Quote
Type:
Lubricants and Oils FT-IR Spectral Library

Thermo Scientific™ Lubricants and Oils FT-IR Spectral Library consists of spectra representing an international collection of lubricants and oils.

This library is an international collection of 1,576 spectra of motor oils, electroinsulating oils, machinery oils, turbine oils, compressor oils, hydraulic oils, anticorrosion oils, steam cylinder oils, metal-cutting oils, mineral oils, synthetic oils, biologically-derived oils, silicone oils, fuels, greases and solvents.
  • Contains a transmission library of over 1000 spectra collected by KBr liquid cell
  • Contains ATR libraries of over 500 spectra in both ATR-corrected and uncorrected formats
  • Data format: High Resolution

Includes:User Guide that provides an overview of lubricant analysis, a description of the sampling techniques used, a list of the most common oil additives, and a table of typical compounds used in synthetic oils and additives and their associated FT-IR absorption bands.

Specifications
For Use With (Application)FT-IR
TypeLubricants and Oils FT-IR Spectral Library
Unit SizeEach